missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentylboronic acid, 95%

Product Code. 11426150
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11426150 10 g 10g
11416150 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11426150 Supplier Thermo Scientific Alfa Aesar Supplier No. H27081.09
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 63076-51-7 | C5H11BO2 | 113.951 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63076-51-7
Molecular Formula C5H11BO2
Molecular Weight (g/mol) 113.951
MDL Number MFCD01074541
InChI Key VTTDFSNKIMAQTB-UHFFFAOYSA-N
Synonym cyclopentaneboronic acid, boronic acid, cyclopentyl, cylopentyl boronic acid, cyclopentylboronicacid, pubchem7792, acmc-209ncu, --paclitaxel;paclitaxel, boronic acid, b-cyclopentyl, cyclopentylboronic acid
PubChem CID 2734327
IUPAC Name cyclopentylboronic acid
SMILES B(C1CCCC1)(O)O

Specifications

Quantity 10 g
Formula Weight 113.95
Percent Purity 95%
Chemical Name or Material Cyclopentylboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.