missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentylboronic acid, 95%, Thermo Scientific™

Product Code. 11935831 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11935831 10 g 10g
11925831 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11935831 Supplier Thermo Scientific Acros Supplier No. 359060100
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 63076-51-7
Molecular Formula C5H11BO2
Molecular Weight (g/mol) 113.95
InChI Key VTTDFSNKIMAQTB-UHFFFAOYSA-N
Synonym cyclopentaneboronic acid, boronic acid, cyclopentyl, cylopentyl boronic acid, cyclopentylboronicacid, pubchem7792, acmc-209ncu, --paclitaxel;paclitaxel, boronic acid, b-cyclopentyl, cyclopentylboronic acid
PubChem CID 2734327
IUPAC Name cyclopentylboronic acid
SMILES B(C1CCCC1)(O)O

Specifications

Melting Point 134.0°C to 142.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 95%
Quantity 10 g
Formula Weight 113.95
Percent Purity 95%
Physical Form Crystalline Powder or Flakes
Chemical Name or Material Cyclopentylboronic acid
compliance-icons
Product Identifier
  • Cyclopentylboronic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.