missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentylmagnesium chloride, 2M solution in diethyl ether, AcroSeal™

Product Code. 10527054 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
100mL
800mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10527054 100 mL AcroSeal™ Glass Bottle 100mL
10071734 800 mL AcroSeal™ Glass Bottle 800mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10527054 Supplier Thermo Scientific Acros Supplier No. 208461000
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 32916-51-1 | C5H9ClMg | 128.88 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32916-51-1 , 60-29-7
Molecular Formula C5H9ClMg
Molecular Weight (g/mol) 128.88
MDL Number MFCD00001357
InChI Key GROGEABZCKQTJW-UHFFFAOYSA-M
Synonym cyclopentylmagnesium chloride, cyclopentyl magnesium chloride, cyclopentylmagnesium chloride solution, cylopentylmagnesium chloride, cyclopentylmagnesium cloride, magnesium,chlorocyclopentyl, cylopentyl magnesium chloride, cyclopentyl-magnesium chloride, grogeabzckqtjw-uhfffaoysa-m, chloro cyclopentyl magnesium
PubChem CID 3599845
IUPAC Name magnesium;cyclopentane;chloride
SMILES Cl[Mg]C1CCCC1

Specifications

Density 0.8600g/mL
Flash Point −40°C
Packaging AcroSeal™ Glass Bottle
Linear Formula C5H9MgCl
Quantity 100 mL
Specific Gravity 0.86
Formula Weight 128.88
Chemical Name or Material Cyclopentylmagnesium chloride, 2M solution in diethyl ether
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cyclopentylmagnesium chloride, 2M solution in diethyl ether
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
  • Substance which, in contact with water, emits flammable gases Category 2
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H261-In contact with water releases flammable gases.
  • H224-Extremely flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P233-Keep container tightly closed.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.