missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Cyclophosphamide

Product Code. 18722554
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18722554 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18722554 Supplier R&D Systems™ Supplier No. 4091/50
  • ONLINE EXCLUSIVE PRICE
    123.80 €
    108.15 € / 50mg
    Save 15.65 €
    12% Off
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Alkylating agent; chemotherapeutic

Nitrogen mustard alkylating agent and prodrug. Phosphoramide mustard (active metabolite) forms DNA cross-links leading to cell death. Inhibits aldehyde dehydrogenase 1 (ALDH1) through its degradation product acrolein. Chemotherapeutic for the treatment of breast cancer; regulates Bax and Bcl-2 expression when administered with etoposide (Cat. No. 1226) in breast cancer cell lines.
TRUSTED_SUSTAINABILITY

Specifications

CAS 50-18-0
Quantity 50 mg
Product Line Tocris
Molecular Formula C7H15Cl2N2O2P
Formula Weight 261.09
Purity >99%
Molecular Weight (g/mol) 261.083
Chemical Name or Material Cyclophosphamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.