missing translation for 'onlineSavingsMsg'
Learn More

Cyclophosphamide monohydrate, 97%

Product Code. 10124460 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10124460 5 g Glass bottle 5g
10751861 1 g Glass bottle 1g
10066333 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10124460 Supplier Thermo Scientific Acros Supplier No. 203960050
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6055-19-2 | C7H17Cl2N2O3P | 279.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6055-19-2
Molecular Formula C7H17Cl2N2O3P
Molecular Weight (g/mol) 279.10
MDL Number MFCD00149395
InChI Key PWOQRKCAHTVFLB-UHFFFAOYNA-N
Synonym cyclophosphamide monohydrate, cyclophosphamide hydrate, cytoxan, endoxon, endoxan monohydrate, ciclophosphamide hydrat, cytoxan hydrate, endoxan a, cyclophosphamide hydrated
PubChem CID 22420
ChEBI CHEBI:4026
IUPAC Name N,N-bis(2-chloroethyl)-2-oxo-1,3,2$l^{5}-oxazaphosphinan-2-amine;hydrate
SMILES O.ClCCN(CCCl)P1(=O)NCCCO1

Specifications

Melting Point 49.0°C to 53.0°C
Color White
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (ELSD) (HPLC)
Packaging Glass bottle
Quantity 5 g
Merck Index 15, 2743
Solubility Information Solubility in water: 40g/L. Other solubilities: slightly soluble in alcohol,benzene,ethylene,glycol,carbon tetrachloride and dioxane,sparingly soluble in ether and acetone
Formula Weight 279.09
Percent Purity 97%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Cyclophosphamide monohydrate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Cyclophosphamide monohydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Carcinogenicity Category 1A
  • Germ cell mutagenicity Category 1B
  • Reproductive toxicity Category 1B
Hazard Statement
  • H350-May cause cancer.
  • H340-May cause genetic defects.
  • H360D-May damage the unborn child.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P264-Wash face, hands and any thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.