missing translation for 'onlineSavingsMsg'
Learn More

Cyclophosphamide Monohydrate, TRC

Product Code. 30133001
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133001 1 g 1g
30134730 5 g 5g
30012129 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133001 Supplier Toronto Research Chemicals Supplier No. TRCC9885801G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6055-19-2
Molecular Formula C7 H15 Cl2 N2 O2 P . H2 O
Molecular Weight (g/mol) 279.1
Synonym 2H-1,3,2-Oxazaphosphorin-2-amine, N,N-bis(2-chloroethyl)tetrahydro-, 2-oxide, monohydrate (9CI), 2H-1,3,2-Oxazaphosphorine, 2-[bis(2-chloroethyl)amino]tetrahydro-, 2-oxide, monohydrate (8CI), 2-[Bis(2-chloroethyl)amino]tetrahydro-2H-1,3,2-oxazaphosphorine-2-oxide monohydrate, Cyclophosphamide hydrate, Cyclophosphamide monohydrate, Endoxan monohydrate
IUPAC Name N,N-bis(2-chloroethyl)-2-oxo-1,3,2λ^{5}-oxazaphosphinan-2-amine;hydrate
SMILES O.ClCCN(CCCl)P1(=O)NCCCO1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C7H15Cl2N2O2P.H2O/c8-2-5-11(6-3-9)14(12)10-4-1-7-13-14;/h1-7H2,(H,10,12);1H2
Formula Weight 278.0354
Percent Purity >95%
Chemical Name or Material Cyclophosphamide Monohydrate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.