missing translation for 'onlineSavingsMsg'
Learn More

Cyclopiazonic Acid, TRC

Product Code. 30137401
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
5 mg
Unit Size:
1mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137401 5 mg 5mg
30010719 2.5 mg 2.5mg
30005543 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137401 Supplier Toronto Research Chemicals Supplier No. TRCC9919955MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18172-33-3
Molecular Formula C20 H20 N2 O3
Molecular Weight (g/mol) 336.38
Synonym 9H-Pyrrolo[1',2':2,3]isoindolo[4,5,6-cd]indol-9-one, 10-acetyl-2,6,6a,7,11a,11b-hexahydro-11-hydroxy-7,7-dimethyl-, (6aα,11aβ,11bα)- (8CI), (6aR,11aS,11bR)-10-Acetyl-2,6,6a,7,11a,11b-hexahydro-11-hydroxy-7,7-dimethyl-9H-pyrrolo[1',2':2,3]isoindolo[4,5,6-cd]indol-9-one, Cyclopiazonic acid, NSC 117181, α-Cyclopiazonic acid, α-Cyclopiazonic Acid, 9H-Pyrrolo[1',2':2,3]isoindolo[4,5,6-cd]indol-9-one, 10-acetyl-2,6,6a,7,11a,11b-hexahydro-11-hydroxy-7,7-dimethyl-, (6aα,11aβ,11bα)- (8CI), (6aR,11aS,11bR)-10-Acetyl-2,6,6a,7,11a,11b-hexahydro-11-hydroxy-7,7-dimethyl-9H-pyrrolo[1',2':2,3]isoindolo[4,5,6-cd]indol-9-one, Cyclopiazonic acid, NSC 117181, α-Cyclopiazonic acid
SMILES CC(=O)C1=C(O)[C@@H]2[C@@H]3[C@@H](Cc4cccc5[nH]cc3c45)C(C)(C)N2C1=O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C20H20N2O3/c1-9(23)14-18(24)17-16-11-8-21-13-6-4-5-10(15(11)13)7-12(16)20(2,3)22(17)19(14)25/h4-6,8,12,16-17,21,24H,7H2,1-3H3/t12-,16+,17+/m1/s1
Formula Weight 336.1474
Chemical Name or Material Cyclopiazonic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.