missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclopropane-d5-carbonyl Chloride, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | Cyclopropanecarbonyl Chloride-D5 |
| CAS | 1219794-96-3 |
| Molecular Formula | C4 D5 Cl O |
| Quantity | 0.5 g |
| Synonym | Cyclopropane-1,2,2,3,3-d5-carbonyl chloride (ACI), Cyclopropane-d5-carbonyl Chloride, Cyclopropanecarbonyl Chloride-D5 |
| InChI Formula | InChI=1 S/C4H5ClO/c5-4(6)3-1-2-3/h3H,1-2H2/i1D2,2D2,3 D |
| SMILES | [2 H]C1([2 H])C([2 H])([2 H])C1([2 H])C(=O)Cl |
| IUPAC Name | 1,2,2,3,3-pentadeuteriocyclopropane-1-carbonyl chloride |
| Molecular Weight (g/mol) | 109.5657 |
| Formula Weight | 109.0343 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction