missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropanecarbonyl Chloride, TRC

Product Code. 30362791
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30362791 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30362791 Supplier Toronto Research Chemicals Supplier No. TRCC9900605G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Spécification

Chemical Name or Material Cyclopropanecarbonyl Chloride
CAS 4023-34-1
Molecular Formula C4H5ClO
Quantity 5 g
Synonym Cyclopropanecarboxylic Acid Chloride, Cyclopropanoyl Chloride, Cyclopropylcarbonyl Chloride, Cyclopropylcarboxylic Acid Chloride
InChI Formula InChI=1S/C4H5ClO/c5-4(6)3-1-2-3/h3H,1-2H2
SMILES ClC(=O)C1CC1
IUPAC Name cyclopropanecarbonyl chloride
Molecular Weight (g/mol) 104.53
Formula Weight 104.0029
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.