missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclopropanecarboxaldehyde, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | Cyclopropanecarboxaldehyde |
| CAS | 1489-69-6 |
| Molecular Formula | C4H6O |
| Quantity | 1 g |
| Synonym | 1-(Formyl)cyclopropane, 2-Cyclopropanecarboxaldehyde, Cyclopropylcarboxaldehyde, Formylcyclopropane |
| InChI Formula | InChI=1S/C4H6O/c5-3-4-1-2-4/h3-4H,1-2H2 |
| SMILES | O=CC1CC1 |
| IUPAC Name | cyclopropanecarbaldehyde |
| Molecular Weight (g/mol) | 70.09 |
| Formula Weight | 70.0419 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction