missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Cyclopropanecarboxamidoxime, 97%

Product Code. 15444558
Change view
Click to view available options
Quantity:
2 g
Unit Size:
2g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15444558 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15444558 Supplier Alfa Aesar™ Supplier No. H58236.04

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 51285-13-3
Molecular Formula C4H8N2O
MDL Number MFCD03426266
Quantity 2 g
Synonym n-hydroxycyclopropanecarboximidamide, z-n'-hydroxycyclopropanecarboxamidine, n-hydroxycyclopropanecarboxamidine, n'-hydroxycyclopropanecarboxamidine, z-n'-hydroxycyclopropanec arboximidamide, cyclopropanecarboxamidoxime, n-hydroxy-cyclopropanecarboximidamide, n'-hydroxycyclopropane-carboximidamide, z-n'-hydroxycyclopropanecarboximidamide, n'-hydroxycyclopropanecarboximideamide
InChI Key OMCUPXRCMTUDHI-UHFFFAOYSA-N
SMILES C1CC1C(=NO)N
IUPAC Name N'-hydroxycyclopropanecarboximidamide
Molecular Weight (g/mol) 100.121
PubChem CID 9582826
Formula Weight 100.12
Percent Purity 97%
Chemical Name or Material Cyclopropanecarboxamidoxime
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.