missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropanecarboxylic acid chloride, 98%

Product Code. 10229783 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10229783 5 g Glass bottle 5g
10010880 25 g Glass bottle 25g
10375980 100 g Glass bottle 100g
11924971 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10229783 Supplier Thermo Scientific Acros Supplier No. 111630050
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4023-34-1 | C4H5ClO | 104.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4023-34-1
Molecular Formula C4H5ClO
Molecular Weight (g/mol) 104.54
InChI Key ZOOSILUVXHVRJE-UHFFFAOYSA-N
Synonym cyclopropanecarboxylic acid chloride, cyclopropane carbonyl chloride, cyclopropylcarboxyl chloride, chloroformyl cyclopropane, cyclopropylcarbonyl chloride, chlorocarbonyl cyclopropane, cyclopropyl carbonyl chloride, cyclopropyl carboxylic chloride, cyclopropane carboxylic acid chloride, cyclopropanoyl chloride
PubChem CID 77637
IUPAC Name cyclopropanecarbonyl chloride
SMILES C1CC1C(=O)Cl

Specifications

Color Colorless to Yellow
Density 1.1500g/mL
Boiling Point 119.0°C
Flash Point 23°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C3H5COCl
Refractive Index 1.4515 to 1.4535
Quantity 5 g
Beilstein 09,4
Specific Gravity 1.15
Solubility Information Solubility in water: decomposes
Formula Weight 104.54
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Cyclopropanecarboxylic acid chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.