missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropanemethylamine, 97%

Product Code. 11491678
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11491678 1 g 1g
11401688 5 g 5g
11411688 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11491678 Supplier Thermo Scientific Alfa Aesar Supplier No. A13946.03
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 2516-47-4 | C4H9N | 71.123 g/mol

Cyclopropanemethylamine used in studies of potential inhibitors of glycine cleavage system of brain and liver mitochondria. It is used in chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopropanemethylamine used in studies of potential inhibitors of glycine cleavage system of brain and liver mitochondria. It is used in chemical synthesis.

Solubility
Fully miscible in water.

Notes
Air sensitive. Store in a cool, dry, well-ventilated area. Store away from oxidizing agent, air , carbon dioxide.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2516-47-4
Molecular Formula C4H9N
Molecular Weight (g/mol) 71.123
MDL Number MFCD00037147
InChI Key IGSKHXTUVXSOMB-UHFFFAOYSA-N
Synonym cyclopropylmethylamine, cyclopropanemethylamine, cyclopropanemethanamine, aminomethylcyclopropane, aminomethyl cyclopropane, 1-cyclopropylmethanamine, cyclopropane methyl amine, cyclopropylmethylamin, cyclopropanmethylamine
PubChem CID 75646
IUPAC Name cyclopropylmethanamine
SMILES C1CC1CN

Specifications

Density 0.82
Boiling Point 85°C to 86°C
Flash Point 2°C (36°F)
Odor Amine-like
Refractive Index 1.433
Quantity 1 g
UN Number UN2733
Beilstein 773674
Sensitivity Air Sensitive
Solubility Information Fully miscible in water.
Formula Weight 71.12
Percent Purity ≥98%
Chemical Name or Material Cyclopropanemethylamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.