missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropanesulfonyl chloride, 97%

Product Code. 15403847
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15403847 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15403847 Supplier Thermo Scientific Alfa Aesar Supplier No. H29107.14
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 139631-62-2 | C3H5ClO2S | 140.581 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 139631-62-2
Density 1.39
Boiling Point 60°C (2 mmHg)
Flash Point 96°C
Molecular Formula C3H5ClO2S
Refractive Index 1.477
MDL Number MFCD01631933
Quantity 25 g
UN Number UN3265
Sensitivity Moisture sensitive
Synonym cyclopropanesulfonylchloride, chlorocyclopropylsulfone, cyclopropanesulphonyl chloride, cyclopropylsulfonyl chloride, cyclopropylsulphonyl chloride
InChI Key PFWWSGFPICCWGU-UHFFFAOYSA-N
SMILES C1CC1S(=O)(=O)Cl
IUPAC Name cyclopropanesulfonyl chloride
Molecular Weight (g/mol) 140.581
PubChem CID 2760919
Formula Weight 140.59
Percent Purity 97%
Chemical Name or Material Cyclopropanesulfonyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.