missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropanethiocarboxamide, 97%

Produktkod. 15485027
Change view
Klicka för att se tillgängliga alternativ
Quantity:
1 g
5 g
Förpackningsstorlek:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Quantity Förpackningsstorlek
15485027 5 g 5g
15475027 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 15485027 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer H32315.06
 Restricted Item
Estimated Shipment: 30-09-2026
Lägg i varukorgen
Lägg i varukorgen
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

CAS: 20295-34-5 | C4H7NS | 101.167 g/mol

Cyclopropanecarbothioamide is used for purification in single-step preparation of thiazolo [5, 4-b] pyridine-and thiazolo [5, 4-c] pyridine derivatives from chloronitropyridines and thioamides, or thioureas.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopropanecarbothioamide is used for purification in single-step preparation of thiazolo [5, 4-b] pyridine-and thiazolo [5, 4-c] pyridine derivatives from chloronitropyridines and thioamides, or thioureas.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 20295-34-5
Molecular Formula C4H7NS
Molecular Weight (g/mol) 101.167
MDL Number MFCD09469287
InChI Key IIPJWNFOLPDTEQ-UHFFFAOYSA-N
Synonym cyclopropanethiocarboxamide, cyclopropanecarbothioic acid amide, cyclopropanethioamide, cyclopropane carbothioamide, cyclopropane-thiocarboxamide, cyclopropane-1-carbothioamide, aminocyclopropylmethane-1-thione
PubChem CID 22140884
IUPAC Name cyclopropanecarbothioamide
SMILES C1CC1C(=S)N

Specifikationer

Melting Point 111°C to 113°C
Quantity 5 g
Formula Weight 101.17
Percent Purity 97%
Chemical Name or Material Cyclopropanethiocarboxamide

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.