missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropyl Isothiocyanate, 97%, Thermo Scientific™

Product Code. 10641685 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
:
1g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10641685 1g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10641685 Supplier Thermo Scientific Acros Supplier No. 375490010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 56601-42-4
Density 1.0700g/mL
Boiling Point 57.0°C to 59.0°C (15.0mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C4H5NS
Linear Formula C3H5NCS
Refractive Index 1.5440 to 1.5460
MDL Number MFCD00040874
Quantity 1g
Specific Gravity 1.07
Synonym cyclopropyl isothiocyanate, cyclopropane, isothiocyanato, isothiocyanato-cyclopropane, cyclopropylisothiocyanate, acmc-20albt, pubchem22785, cyclopropyl iso-thiocyanate, cyclopropane,isothiocyanato
InChI Key JGFBQFKZKSSODQ-UHFFFAOYSA-N
SMILES S=C=NC1CC1
IUPAC Name isothiocyanatocyclopropane
Molecular Weight (g/mol) 99.15
PubChem CID 92463
Formula Weight 99.16
Percent Purity 97%
Chemical Name or Material Cyclopropyl isothiocyanate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cyclopropyl isothiocyanate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H301-Toxic if swallowed.
  • H331-Toxic if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.