missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropyl isothiocyanate, 97%

Product Code. 11328545
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11328545 1 g 1g
11338545 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 11328545

Brand: Thermo Scientific Alfa Aesar L11985.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 56601-42-4 | C4H5NS | 99.15 g/mol

Cyclopropyl isothiocyanate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopropyl isothiocyanate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56601-42-4
Molecular Formula C4H5NS
Molecular Weight (g/mol) 99.15
MDL Number MFCD00040874
InChI Key JGFBQFKZKSSODQ-UHFFFAOYSA-N
Synonym cyclopropyl isothiocyanate, cyclopropane, isothiocyanato, isothiocyanato-cyclopropane, cyclopropylisothiocyanate, acmc-20albt, pubchem22785, cyclopropyl iso-thiocyanate, cyclopropane,isothiocyanato
PubChem CID 92463
IUPAC Name isothiocyanatocyclopropane
SMILES S=C=NC1CC1

Specifications

Density 1.07
Boiling Point 87°C to 88°C (35 mmHg)
Flash Point 57°C (134°F)
Refractive Index 1.548
Quantity 1 g
UN Number UN2929
Beilstein 3600974
Sensitivity Moisture sensitive
Formula Weight 99.16
Percent Purity 97%
Chemical Name or Material Cyclopropyl isothiocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.