missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropyl methyl ketone, 98%

Product Code. 13488308 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
500mL
This item is not returnable. View return policy

Product Code. 13488308

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 765-43-5 | C5H8O | 84.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 765-43-5
Molecular Formula C5H8O
Molecular Weight (g/mol) 84.12
MDL Number MFCD00001297
InChI Key HVCFCNAITDHQFX-UHFFFAOYSA-N
Synonym cyclopropyl methyl ketone, acetylcyclopropane, methyl cyclopropyl ketone, ethanone, 1-cyclopropyl, 1-cyclopropyl-ethanone, 1-cyclopropylethan-1-one, ketone, cyclopropyl methyl, cyclopropylmethylketone, unii-n27yy1xcfh, cyclopropylethanone
PubChem CID 13004
IUPAC Name 1-cyclopropylethanone
SMILES CC(=O)C1CC1

Specifications

Melting Point -68.0°C
Color Colorless to Yellow
Density 0.8980g/mL
Boiling Point 111.0°C
Flash Point 21°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C3H5COCH3
Refractive Index 1.4220 to 1.4260
Quantity 500 mL
Beilstein 07,7
Fieser 01,676
Specific Gravity 0.898
Solubility Information Solubility in water: 185g/L (20°C). Other solubilities: soluble in alcohols,chloride solvents
Viscosity 0.65 mPa.s (20°C)
Formula Weight 84.12
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Cyclopropyl methyl ketone
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Cyclopropyl methyl ketone
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.