missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropyl(phenyl)sulfane, 98%, BLD Pharm™

Product Code. 30775332
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Unit Size:
1g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30775332 1 g 1g
30767510 25 g 25g
30767408 100 g -
30768390 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 30775332 Supplier BLD Pharm Supplier No. BD1230431g
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 14633-54-6
Molecular Formula C9H10S
MDL Number MFCD00009684
Quantity 1 g
SMILES C1(SC2CC2)=CC=CC=C1
Molecular Weight (g/mol) 150.24
Percent Purity 98%
Chemical Name or Material Cyclopropyl(phenyl)sulfane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.