missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropylboronic acid, 90%, Thermo Scientific™

Product Code. 10747105
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10747105 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10747105 Supplier Thermo Scientific Maybridge Supplier No. MAY00065DA
 Restricted Item
Estimated Shipment: 13-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 411235-57-9
CAS Min % 90.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Molecular Formula C3H7BO2
Linear Formula C3H5B(OH)2
MDL Number MFCD04038750
Quantity 1 g
Synonym cyclopropyl boronic acid, cyclopropylboranediol, cyclopropaneboronic acid, boronic acid, cyclopropyl, cyclopropyl-boronic acid, boronic acid, cyclopropyl-9ci, cyclopropylboronicacid, boronocyclopropane, cyclopropyboronic acid, cyclopropylboromc acid
InChI Key WLVKDFJTYKELLQ-UHFFFAOYSA-N
SMILES OB(O)C1CC1
IUPAC Name cyclopropylboronic acid
Molecular Weight (g/mol) 85.90
PubChem CID 2760897
Formula Weight 85.9
Percent Purity 90%
Chemical Name or Material Cyclopropylboronic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.