missing translation for 'onlineSavingsMsg'
Learn More

cyclopropylboronic acid, 97%, Thermo Scientific™

Product Code. 10672685
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10672685 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10672685 Supplier Thermo Scientific Maybridge Supplier No. MAY00010DA
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 126689-01-8
CAS Min % 97.0
Density 0.9220g/mL
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C9H17BO2
MDL Number MFCD05663847
Quantity 1 g
Synonym cyclopropylboronic acid pinacol ester, cyclopropylboronic acid, pinacol ester, 4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl cyclopropane, 1,3,2-dioxaborolane, 2-cyclopropyl-4,4,5,5-tetramethyl, 2-cyclopropyl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane, 2-cyclopropyl-4,4,5,5-tetramethyl-1,3,2-dioxaborol, cypbe, pubchem4015, pubchem7931, acmc-209bbh
InChI Key XGBMQBPLWXTEPM-UHFFFAOYSA-N
SMILES CC1(C)OB(OC1(C)C)C1CC1
IUPAC Name 2-cyclopropyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol) 168.04
PubChem CID 2758015
Formula Weight 168.04
Percent Purity 97%
Chemical Name or Material Cyclopropylboronic acid pinacol ester
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.