missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropylmagnesium bromide, 0.5M solution in THF, AcroSeal™, Thermo Scientific Chemicals

Product Code. 11376759
Change view
Click to view available options
Quantity:
25 mL
100 mL
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11376759 100 mL 100mL
11366759 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11376759 Supplier Thermo Scientific Alfa Aesar Supplier No. H26273.AE
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 23719-80-4 | Molecular Formula: C3H5BrMg | Molecular Weight: 145.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It often acts as the starting nutrient mix for dropout studies where fermentation conditions required the absence of one or more essential amino acids. For classifying Yeasts based on colonial characteristics and cell morphology.

Solubility
Soluble in water (17 mg/ml), (25°C). Soluble in organic solvents.

Notes
Air & Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air, water and moisture.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23719-80-4
Molecular Formula C3H5BrMg
Molecular Weight (g/mol) 145.28
MDL Number MFCD00672094
InChI Key QUPLLJGAJVXXBW-UHFFFAOYSA-M
Synonym cyclopropylmagnesium bromide, cyclopropylmagnesiumbromide, bromo cyclopropyl magnesium, magnesium, bromocyclopropyl, cyclopropyl magnesium bromide, cprmgbr, cyclopropyl magnesiumbromide, magnesium,bromocyclopropyl, bromo-cyclopropyl magnesium, cyclopropylmagnesiunn bromide, grignard reagent
PubChem CID 11116189
IUPAC Name magnesium;cyclopropane;bromide
SMILES Br[Mg]C1CC1

Specifications

Density 0.968
Flash Point −17°C (1°F)
Quantity 100 mL
UN Number UN2924
Sensitivity Air and moisture sensitive; May form a precipitate
Solubility Information Soluble in water (17mg/ml),(25°C). Soluble in organic solvents.
Formula Weight 145.28
Concentration or Composition (by Analyte or Components) 0.5 to 0.7M in THF
Chemical Name or Material Cyclopropylmagnesium bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.