missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropylmethyl Bromide, TRC

Product Code. 30354006
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30354006 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30354006 Supplier Toronto Research Chemicals Supplier No. TRCC98872510G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cyclopropylmethyl Bromide
CAS 7051-34-5
Molecular Formula C4H7Br
Quantity 10 g
Synonym (Bromomethyl)cyclopropane, 1-(Bromomethyl)cyclopropane, Cyclopropanemethyl Bromide, Cyclopropylcarbinyl Bromide
InChI Formula InChI=1S/C4H7Br/c5-3-4-1-2-4/h4H,1-3H2
SMILES C1CC1CBr
IUPAC Name bromomethylcyclopropane
Molecular Weight (g/mol) 135
Formula Weight 133.9731
Name des Produkts
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.