missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles

Product Code. 15435305
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15435305 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15435305 Supplier Thermo Scientific Alfa Aesar Supplier No. H58008.AD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 126403-68-7 | C3H5BrZn

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopropylzinc bromide acts as an active nucleophile in palladium-catalyzed cross-coupling of heteroaryl halides.

Solubility
Miscible with tetrahydrofuran.

Notes
Heat and moisture sensitive. Store
TRUSTED_SUSTAINABILITY

Specifications

CAS 126403-68-7
Density 0.969
Molecular Formula C3H5BrZn
MDL Number MFCD08741505
Quantity 50 mL
UN Number UN3399
Sensitivity Air sensitive
Solubility Information Miscible with tetrahydrofuran.
Formula Weight 186.36
Chemical Name or Material Cyclopropylzinc bromide, Packaged under Argon in resealable ChemSeal™ bottles

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.