missing translation for 'onlineSavingsMsg'
Learn More

TOKU-E™ Cyclosporin A ReadyMade™ Solution

Product Code. 30044045
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 1 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30044045 10 x 1 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30044045 Supplier TOKU-E™ Supplier No. C28510x1ML
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Cyclosporin A ReadyMade™ Solution is provided as a sterile-filtered solution of Cyclosporin A in DMSO.

  • CAS Number: 59865-13-3
  • Molecular Formula: C62H111N11O12
  • Molecular Weight: 1202.61 g/mol
  • Storage Conditions: -20°C
  • Form: Liquid
  • Source: Tolypocladium inflatum
  • Spectrum: Cyclosporin A is a calcineurin inhibitor with immunosuppressive properties. It can be used in organ-on-a-chip models to study nephrotoxicity and is also being studied for utility for SARS-CoV-2
  • Mechanism of Action: Cyclosporin A can inhibit function of many nuclear proteins involved in T-cell activation at the level of mRNA transcription. It forms a complex with cyclophilin, which can bind to calcineurin, a Ca2+- and calmodulin-dependent protein phosphatase, inhibiting its enzymatic activity

Specifications

CAS 59865-13-3
Certifications/Compliance Purity >99%
Chemical Name or Material Cyclosporin A
Color Clear
Content And Storage -20°C, Protect from light
Description Cyclosporin A ReadyMade™ Solution 5 mg/mL in DMSO
Concentration 5 mg/mL
Detection Method HPLC
Form Clear, colorless solution
Product Type Antibiotic
Sterility Sterile
Sterilization Method Sterile-filtered
Formulation Liquid in DMSO
For Use With (Application) Microbiology, Eukaryotic Cell Culture Application
Product Line ReadyMade™ Solutions
Purity or Quality Grade >99%
Quantity 10 x 1 mL
Shipping Condition Ice Pack
Source Tolypocladium inflatum
Species Bacteria
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.