missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Cyclosporin A

Product Code. 18762494
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18762494 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18762494 Supplier Tocris Bioscience™ Supplier No. 1101/100
  • ONLINE EXCLUSIVE PRICE
    217.00 €
    204.75 € / 100mg
    Save 12.25 €
    5% Off
 Restricted Item
Estimated Shipment: 27-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Calcineurin inhibitor

Immunosuppressant; inhibits phosphatase activity of calcineurin (IC50 = 5 nM). Also inhibits formation and opening of the mitochondrial permeability transition pore (MPTP).
TRUSTED_SUSTAINABILITY

Specifications

CAS 59865-13-3
Quantity 100 mg
Molecular Formula C62H111N11O12
Formula Weight 1202.63
Purity >99%
Molecular Weight (g/mol) 1202.64
Chemical Name or Material Cyclosporin A
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.