missing translation for 'onlineSavingsMsg'
Learn More

Cyclosporin A, TRC

Product Code. 30363294
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30363294 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30363294 Supplier Toronto Research Chemicals Supplier No. TRCC9889001G
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cyclosporin A
CAS 59865-13-3
Purity Grade Notes HPLC
Molecular Formula C62 H111 N11 O12
Quantity 1 g
Synonym 1,4,7,10,13,16,19,22,25,28,31-Undecaazacyclotritriacontane, cyclic peptide deriv., 1: PN: US6686454 SEQID: 1 claimed protein, 7: PN: WO2000002548 PAGE: 30 claimed protein, Abrammune, Antibiotic S 7481F1, Arpimune ME, CICPORAL, Cicloral, Cicloral (antibiotic), Ciclosporin, Ciclosporina Germed, Cipol N, Ciqorin, Consupren, CyclASol, Cyclo[L-alanyl-D-alanyl-N-methyl-L-leucyl-N-methyl-L-leucyl-N-methyl-L-valyl-(3R,4R,6E)-6,7-didehydro-3-hydroxy-N,4-dimethyl-L-2-aminooctanoyl-L-2-aminobutanoyl-N-methylglycyl-N-methyl-L-leucyl-L-valyl-N-methyl-L-leucyl], Cyclokat, Cyclosporin, Cyclosporine, Cyclosporine A, Cypol-N, Debio088, Equoral, Gengraf, Imusporin, NSC 290193, Neoplanta, Neoral, OL 27-400, Optimmune, Papilock Mini, Ramihyphin A, Restasis, S-Neoral, SDZ-OXL 400, Sandimmun, Sandimmun Neoral, Sandimmun Optoral, Sandimmune, Sandimmune Neoral, Sang-35, SangCyA, Sigmasporin Microoral, Zinograf ME, Ciclosporin, Cyclo[[(2S,3R,4R,6E)-3-hydroxy-4-methyl-2-(methylamino)oct-6-enoyl]-l-2-aminobutanoyl-N-methylglycyl-N-methyl-l-leucyl-l-valyl-N-methyl-l-leucyl-l-alanyl-d-alanyl-N-methyl-l-leucyl-N-methyl-l-leucyl-N-methyl-l-valyl], ciclosporin A, 1,4,7,10,13,16,19,22,25,28,31-Undecaazacyclotritriacontane, cyclic peptide deriv., Abrammune, Antibiotic S 7481F1, Arpimune ME, CICPORAL, Cicloral, Cicloral (antibiotic), Ciclosporina Germed, Cipol N, Consupren, Cyclo[L-alanyl-D-alanyl-N-methyl-L-leucyl-N-methyl-L-leucyl-N-methyl-L-valyl-(3R,4R,6E)-6,7-didehydro-3-hydroxy-N,4-dimethyl-L-2-aminooctanoyl-L-2-aminobutanoyl-N-methylglycyl-N-methyl-L-leucyl-L-valyl-N-methyl-L-leucyl], Cyclokat, Cyclosporin, Cyclosporine, Cyclosporine A, Debio088, Equoral, Gengraf, Imusporin, NSC 290193, Neoplanta, Neoral, OL 27-400, Optimmune, Papilock Mini, Ramihyphin A, Restasis, S-Neoral, SDZ-OXL 400, Sandimmun, Sandimmun Neoral, Sandimmun Optoral, Sandimmune, Sandimmune Neoral, Sang-35, SangCyA, Sigmasporin Microoral, Zinograf ME
InChI Formula InChI=1S/C62H111N11O12/c1-25-27-28-40(15)52(75)51-56(79)65-43(26-2)58(81)67(18)33-48(74)68(19)44(29-34(3)4)55(78)66-49(38(11)12)61(84)69(20)45(30-35(5)6)54(77)63-41(16)53(76)64-42(17)57(80)70(21)46(31-36(7)8)59(82)71(22)47(32-37(9)10)60(83)72(23)50(39(13)14)62(85)73(51)24/h25,27,34-47,49-52,75H,26,28-33H2,1-24H3,(H,63,77)(H,64,76)(H,65,79)(H,66,78)/b27-25+/t40-,41+,42-,43+,44+,45+,46+,47+,49+,50+,51+,52-/m1/s1
SMILES CC[C@@H]1NC(=O)[C@H]([C@H](O)[C@H](C)C\C=C\C)N(C)C(=O)[C@H](C(C)C)N(C)C(=O)[C@H](CC(C)C)N(C)C(=O)[C@H](CC(C)C)N(C)C(=O)[C@@H](C)NC(=O)[C@H](C)NC(=O)[C@H](CC(C)C)N(C)C(=O)[C@@H](NC(=O)[C@H](CC(C)C)N(C)C(=O)CN(C)C1=O)C(C)C
IUPAC Name (3S,6S,9S,12R,15S,18S,21S,24S,30S,33S)-30-ethyl-33-[(E,1R,2R)-1-hydroxy-2-methylhex-4-enyl]-1,4,7,10,12,15,19,25,28-nonamethyl-6,9,18,24-tetrakis(2-methylpropyl)-3,21-di(propan-2-yl)-1,4,7,10,13,16,19,22,25,28,31-undecazacyclotritriacontane-2,5,8,11,14,17,20,23,26,29,32-undecone
Molecular Weight (g/mol) 1202.61
Formula Weight 1201.8414
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.