missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Cyclosporin H

Product Code. 18799206
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18799206 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18799206 Supplier Tocris Bioscience™ Supplier No. 6982/5
  • ONLINE EXCLUSIVE PRICE
    426.00 €
    402.15 € / 5mg
    Save 23.85 €
    5% Off
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Enhances lentiviral transduction

Cyclosporin H is a viral transduction enhancer. Increases lentiviral transduction up to 10-fold in human cord blood-derived hematopoietic stem and progenitor cells (HSPCs). Displays an additive effect when combined with rapamycin (Cat. No. 1292) or prostaglandin E2 (Cat. No. 2296). Lacks immunosuppressant activity of cyclosporin A (Cat. No. 1101).
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-(N-Methyl-D-valine)cyclosporin A
CAS 83602-39-5
Quantity 5 mg
Target Viral Transduction Enhancers
Molecular Formula C62H111N11O12
Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.