missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Cyclothiazide

Product Code. 30001116
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30001116 10 mg 10mg
18722674 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30001116 Supplier R&D Systems™ Supplier No. 0713/10
  • ONLINE EXCLUSIVE PRICE
    157.00 €
    148.05 € / 10mg
    Save 8.95 €
    5% Off
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Positive allosteric modulator of AMPA receptors; inhibits AMPA desensitization

Positive allosteric modulator of AMPA receptors that potently inhibits AMPA receptor desensitization. Selective for the flip variant of each of the four receptor subunits.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2259-96-3
Molecular Formula C14H16ClN3O4S2

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 389.87
Purity >98%
Molecular Weight (g/mol) 389.869
Chemical Name or Material Cyclothiazide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.