missing translation for 'onlineSavingsMsg'
Learn More

Cytiva CyDye™ DIGE Fluors: Cy5

Product Code. 10269114 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
2 x 5 nM
5 nM
5 x 5 nM
Unit Size:
Each
Product Code. Quantity unitSize
10269114 5 x 5 nM Each
10116624 2 x 5 nM Each
10407104 5 nM Each
3 options
This item is not returnable. View return policy

Product Code. 10269114

Brand: Cytiva RPK0275

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Allow detection of up to three prelabeled protein samples and standards on the same 2-D electrophoresis gel

  • Size- and charge-matched dyes enable co-migration of labeled samples within the gel
  • Bright and highly sensitive dyes allow the use of the minimal labeling technique
  • Minimal loss of signal during labeling, separation, and scanning
  • No change in signal over wide pH range used during first-dimension (IEF) separation
  • Discrete signal from each fluor with minimal cross-talk contributes to high accuracy
  • Allow the labeling of 50μg protein per labeling reaction

For general 2-D applications where sufficient amounts of sample are available.

TRUSTED_SUSTAINABILITY

Specifications

Format Solid
For Use With (Application) For general 2-D applications where sufficient amounts of sample are available, detection of up to three prelabeled protein samples and standards on the same 2-D electrophoresis gel
Quantity 5 x 5 nM
Product Type Amersham CyDye DIGE Fluor Cy5 Minimal Dye
Kit Contents 25nM of CyDye DIGE Fluor Cy5 minimal dye
Sufficient For Larger experimental set-ups with several variables, up to 60 labeling
Content And Storage -15°C to -30°C
Color Red
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H317-May cause an allergic skin reaction.
  • H332-Harmful if inhaled.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P501-Dispose of contents/container to .
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.