missing translation for 'onlineSavingsMsg'
Learn More

KGW Isotherm™ Cylindrical Dewar Flasks, Blue with Handle

Product Code. 15781977
Click to view available options
Capacity (Metric):
1 L
1500 mL
Diameter (Metric) Outer:
95-105 mm
120-130 mm
Height (Metric):
290 mm
300 mm
Model:
G 15 C
G 9 C
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15781977

Brand: KGW Isotherm™ 1068

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Dewar flasks according to DIN EN ISO 16496. KGW-Isotherm Cylindrical Dewar Flasks, Blue with Side Handle provide high insulation reliability and easy handling in an economical product. Available in a variety of capacities, with plugs sold separately.

  • Made of borosilicate glass according to ISO 3585
  • Protective casing made of blue-coated metal
  • Fully silvered
  • Easy handling with side grip handle
  • Area of application:
    • Flexible laboratory, medical and biotechnology applications
    • Insulated storing and short-distance transport of small samples
    • Store under liquid or solid coolants (e.g. liquid N2 and CO2 dry ice)
    • Pressure-less space for coolants
    Chemical characteristics:
    • Hydrolytic resistance: according to ISO 719 (98°C)
    • Hydrolytic resistance: according to ISO 720 (121°C)
    • Acid resistance: according to ISO 1776
    • Alkaline resistance: according to ISO 695-A2
  • Temperature range:
    • Glass refill from -200°C to + 150°C, for a short time 200°C
    • Casing with mounting rubber and Latizell buffer from -10°C to + 60°C

Specifications

Cap Lid Type Plug
Color Blue
Diameter (Metric) Outer 95-105 mm
For Use With (Application) Insulated storage and small distance transport
For Use With (Equipment) Coolant
Height (Metric) Inner 235 mm
Height (Metric) 290 mm
Includes Handle/Side grip
Model G 9 C
Base Material Coated Metal
Capacity (Metric) 1 L
Diameter (Metric) Inner 77 mm
Material Borosilicate Glass
Style Fully silvered
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.