missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ CYM 5520

Product Code. 18084972
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18084972

Brand: Tocris Bioscience™ 5418/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective S1P2 allosteric agonist

CYM 5520 is a selective S1P2 allosteric agonist (EC50 = 0.48 μM). Selective for S1P2 receptors over a panel of 29 other receptors and transporters.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-[2-[2,5-Dimethyl-1-(phenylmethyl)-1H-pyrrol-3-yl]-2-oxoethyl]-1,6-dihydro-6-oxo-3-pyridinecarbonitrile
CAS 1449747-00-5
Quantity 10 mg
Target Sphingosine-1-phosphate Receptor Agonists
Molecular Formula C21H19N3O2
Purity 0.99
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.