Learn More
Tocris Bioscience™ CYN 154806
GREENER_CHOICE
Selective sst2 antagonist
Brand:  Tocris Bioscience™ 1843/1
Encompass
Product Code. 13490104
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
CYN 154806 is a potent and selective somatostatin sst2 receptor antagonist. pIC50 values are 8.58, 5.41, 6.07, 5.76 and 6.48 for human recombinant sst2, sst1, sst3, sst4 and sst5 receptors respectively.
Specifications
Specifications
95%
CYN 154806
CYN 154806
Ac-(4-Nitro)Phe-D-Cys-Tyr-D-Trp-Lys-Thr-Cys-D-Tyr-NH2 (Disulfide bridge: 2 - 7)
Store at -20°C
1197.35