missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Cynomolgus Monkey IDUA (XP_005554313.1) VersaClone cDNA

Product Code. 18421710
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18421710 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18421710 Supplier R&D Systems™ Supplier No. RDC3448
  • ONLINE EXCLUSIVE PRICE
    761.00 €
    719.25 € / 10µg
    Save 41.75 €
    5% Off
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Contains gene inserts that are fully sequenced and produced with high quality

TRUSTED_SUSTAINABILITY

Specifications

Gene alpha-L-Iduronidase/IDUA
Accession Number XP_005554313.1
Gene ID (Entrez) 3425
Gene Alias alphaLIduronidase, IDA, IDUA, MPS1, MPSI
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Cynomolgus Monkey
Product Line R&D Systems
Quantity 10 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.