missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Cynomolgus Monkey ITGB2/CD18 (XP_005548577) VersaClone cDNA

Product Code. 18708669
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18708669 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18708669 Supplier R&D Systems™ Supplier No. RDC2707
  • ONLINE EXCLUSIVE PRICE
    341.00 €
    322.35 € / 10µg
    Save 18.65 €
    5% Off
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This shuttle vector contains the complete ORF for the gene of interest, along with a Kozak consensus sequence for optimal translation initiation. It is inserted NotI to AscI. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector.

  • Integrins are integral cellsurface proteins composed of an alpha chain and a beta chain.
  • They are known to participate in cell adhesion as well as cellsurface mediated signaling.
  • ITGB2 belongs to the integrin beta chain family of proteins.
  • It combines with the alpha L chain to form the integrin LFA-1 and combines with the alpha M chain to form the integrin Mac1.
  • Defects in ITGB2 are the cause of leukocyte adhesion deficiency type I (LAD1).
  • The insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
  • For reasearch use only. Not for use in humans.
TRUSTED_SUSTAINABILITY

Specifications

Gene cynoITGB2
Accession Number XP_005548577
Concentration 10 μg at 0.2 μg/μL
Formulation cDNA is provided in 10 mM Tris-Cl, pH 8.5
Insert Type 2323 bp
Gene ID (Entrez) 3689
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Cynomolgus Monkey
Quantity 10 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.