missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Cynomolgus Monkey PSMA/FOLH1 (XP_005579379) VersaClone cDNA
Specifications
Specifications
| Gene | PSMA/FOLH1/NAALADase I |
| Accession Number | XP_005579379 |
| Gene ID (Entrez) | 2346 |
| Gene Alias | Cell growth-inhibiting gene 27 protein, cell growth-inhibiting protein 27, EC 3.4.17.21, FGCP, folate hydrolase (prostate-specific membrane antigen) 1, Folate hydrolase 1, FOLH1, FOLHNAALADase I, Folylpoly-gamma-glutamate carboxypeptidase, GCP2, GCPII, GCPIImGCP, glutamate carboxylase II, glutamate carboxypeptidase 2, Glutamate carboxypeptidase II, Membrane glutamate carboxypeptidase, mopsm, NAALAD1, NAALAD1N-acetylated-alpha-linked acidic dipeptidase I, NAALAdase, NAALADase I, N-acetylated alpha-linked acidic dipeptidase 1, prostate specific membrane antigen variant F, Prostate-specific membrane antigen, PSMAFGCP, PSMPteroylpoly-gamma-glutamate carboxypeptidase |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Cynomolgus Monkey |
| Quantity | 10 μg |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?