Learn More
Riley™ Cypher Eco Safety Glasses
Product Code. 31702885
Change view
Click to view available options
Lens Tint:
Blue
Clear
Gray
LED
Unit Size:
Pack of 20
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Lens Tint Unit Size
31702885 Clear Pack of 20
31702884 Gray Pack of 20
31702882 Blue Pack of 20
31702883 LED Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 31702885 Supplier Riley™ Supplier No. RLY00611
  • ONLINE EXCLUSIVE PRICE
    615.50 €
    208.13 € / Pack of 20
    Save 407.37 €
    66% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Sleek safety glasses with a curved frame and soft temples for enhanced comfort, offering reliable protection and eco-friendly design.
  • Sports style 8 Base lens
  • Soft, adjustable anti-slip nose bridge
  • Co-injected bi-colour temples with flexible TPE ends enhance overall fit and comfort
  • No single-use packaging, supplied in a microfiber pouch made from 100% recycled material
  • Unique aero-slotted side arms
  • Available for prescription
Specifications
Certifications/Compliance CE or - UKCA or - EN 166:2001
Lens Coating Anti-fog, Scratch-resistant, Moisture Repellent, UV-treated
Lens Tint Clear
Product Type Safety Glasses
For Use With (Application) Component and Product Assembly, Vehicle Repairs and Maintenance, Commercial Cleaning, General Maintenance, Inspection and Quality Control
Frame Color Black
Transmittance Value 92%
Weight (Metric) 22 g
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate (High-impact)
Frame Material Polycarbonate
UV Protection UV400
Frame Type Frameless
Optical Clarity 2C
Mechanical Strength F,T