missing translation for 'onlineSavingsMsg'
Learn More

Cytiva CypHer5E NHS Esters

Product Code. 11505834 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
Each
Product Code. Quantity unitSize
11505834 1 mg 1mg
11515834 5 mg Each
2 options
This item is not returnable. View return policy

Product Code. 11505834

Brand: Cytiva PA15401

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A red-excitable, pH-sensitive cyanine dye derivative, reporting receptors movement from the cell surface into acidic endosomes upon agonist stimulation.

CypHer5E is a flexible, generic fluorophore that can be used to study most classes of cell surface receptors which internalize into the endocytic pathway when activated by a ligand.

  • Generic technology can be coupled to any receptor or epitope tag antibody.
  • Monitors any neutral to acidic event.
  • Simple fluorescence intensity increase.
  • Classic CyDye profile of two heterocycles linked by a polymethine bridge, and contains secondary nitrogen atoms in one or both heterocyclic rings.
  • CypHer5E has additional sulfonate groups to enhance its solubility in aqueous solutions.
  • Increased labelling efficiency and reduces adverse dye-dye interactions.

CypHer5E reports ligand-mediated receptor internalization by exploiting pH differences that occur along the endocytic pathway. CypHer5E is a red-excitable, pH-sensitive cyanine dye derivative that is minimally fluorescent at a basic pH and maximally fluorescent at an acidic pH. It is therefore well suited to report the movement of a receptor from the cell surface into acidic endosomes upon agonist stimulation.

TRUSTED_SUSTAINABILITY

Specifications

Formulation 100% CypHer 5E
For Use With (Application) This reagent has been shown to be suitable for live cell receptor internalization studies using the IN Cell Analyzer 3000, IN Cell Analyzer 1000, and confocal microscopes. 0.5 mL of stock reagent will be sufficient for up to 500 wells.
Quantity 1 mg
Product Type CypHer5E NHS Ester
Sufficient For 50Wells
Solubility Easily soluble in methanol; Very slightly soluble in cold water and hot water.
Label or Dye Cy
Color Blue
Emission 663
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin sensitiser Category 1
  • Respiratory sensitiser Category 1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.