Learn More
Description
CypHer5E is a flexible, generic fluorophore that can be used to study most classes of cell surface receptors which internalize into the endocytic pathway when activated by a ligand.
- Generic technology can be coupled to any receptor or epitope tag antibody.
- Monitors any neutral to acidic event.
- Simple fluorescence intensity increase.
- Classic CyDye profile of two heterocycles linked by a polymethine bridge, and contains secondary nitrogen atoms in one or both heterocyclic rings.
- CypHer5E has additional sulfonate groups to enhance its solubility in aqueous solutions.
- Increased labelling efficiency and reduces adverse dye-dye interactions.
CypHer5E reports ligand-mediated receptor internalization by exploiting pH differences that occur along the endocytic pathway. CypHer5E is a red-excitable, pH-sensitive cyanine dye derivative that is minimally fluorescent at a basic pH and maximally fluorescent at an acidic pH. It is therefore well suited to report the movement of a receptor from the cell surface into acidic endosomes upon agonist stimulation.
Specifications
Specifications
| Formulation | 100% CypHer 5E |
| For Use With (Application) | This reagent has been shown to be suitable for live cell receptor internalization studies using the IN Cell Analyzer 3000, IN Cell Analyzer 1000, and confocal microscopes. 0.5 mL of stock reagent will be sufficient for up to 500 wells. |
| Quantity | 5 mg |
| Product Type | CypHer5E NHS Ester |
| Solubility | Easily soluble in methanol; Very slightly soluble in cold water and hot water. |
| Color | Blue |
| Emission | 663 |
Safety and Handling
- Danger
- Acute toxicity Category 4
- Skin sensitiser Category 1
- Respiratory sensitiser Category 1
- H302 + H312 + H332-Harmful if swallowed, in contact with skin or if inhaled.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H317-May cause an allergic skin reaction.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- P501-Dispose of contents/container to .
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.