missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ pTK-Cypridina Luc Vector for Luciferase Assays

Product Code. 12152230 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12152230 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12152230 Supplier Thermo Scientific™ Supplier No. 16151
  • ONLINE EXCLUSIVE PRICE
    137.00 €
    110.97 € / 10µg
    Save 26.03 €
    19% Off
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Drive Cypridina luciferase expression as a reporter for gene regulation studies using vectors with a multiple cloning site (MCS) or control promoter (CMV or TK).

The Thermo Scientific™ pMCS-Cypridina Luc Vector is a multiple cloning site plasmid designed to accept a promoter sequence for study of gene regulation using the naturally secreting Cypridina luciferase reporter.

The Cypridina Luc Vectors contain a gene cloned from the marine ostracod, Cypridina noctiluca. The gene encodes a naturally-secreted bioluminescent Cypridina luciferase (62kDa), which enables measurement of the reporter activity in media (for real-time assays) and in cell lysates. The pMCS vector contains a multiple cloning site for cloning a promoter to study its regulatory potential. The pCMV and pTK vectors have the luciferase gene under the CMV (Cytomegalovirus) promoter and Herpes Simplex Virus (HSV) thymidine kinase (TK) promoter, respectively. These constitutive expression vectors can be used as normalization controls to account for experimental variation in combination with other reporters.

Highlights:

  • Naturally-secreting Cypridina luciferase gene, optimized for high expression in mammalian systems
  • Multiple cloning site (MCS) provides versatility for transfer of regulatory elements from one plasmid to another
  • Transcription termination site (Ter), Lac operator (Lac O1), and transcriptional pause site (TPS) used to minimize background by reducing transcriptional read-through
  • Both puromycin (Pur) and ampicillin (Amp) markers for drug selection in mammalian and bacterial cells, respectively
  • High-copy pUC bacterial DNA replication origin
  • Two control vectors available with strong (CMV) or weak (TK) constitutive promoters for co-transfection and normalization
TRUSTED_SUSTAINABILITY

Specifications

Description pTK-Cypridina Luc Vector
Type Luc Vector
Cloning Method Restriction Enzyme/MCS
Delivery Type Transfection
Quantity 10 μg
Content And Storage Store between -70°C and -20°C.
Reporter Gene Cypridina Luciferase
Selection Agent (Eukaryotic) Puromycin
Product Line Pierce
For Use With (Application) Reporter Assays
Promoter TK
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.