missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals [Cys8] Renin Substrate Tetradecapeptide, Rat

Product Code. 15477279
Change view
Click to view available options
Substrate Quantity:
1mg
5mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Substrate Quantity Unit Size
15477279 5mg 5mg
15487279 1mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15477279 Supplier Thermo Scientific Chemicals Supplier No. J66067.#0
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For cellular and molecular biology applications

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Spécification

Color White to off-white
Form Solid
Molecular Weight (g/mol) 1779.05
Storage Requirements -30° to -10°C
Substrate Quantity 5mg

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.