missing translation for 'onlineSavingsMsg'
Learn More

Cystamine dihydrochloride, min. 97.0%, MP Biomedicals™

Product Code. 11447540 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11447540 1 g 1g
11457540 5 g 5g
11467540 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11447540 Supplier MP Biomedicals Supplier No. 0210049201
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 56-17-7
Molecular Formula C4H14Cl2N2S2
Molecular Weight (g/mol) 225.19
InChI Key YUFRRMZSSPQMOS-UHFFFAOYSA-N
Synonym cystamine dihydrochloride, cystamin dihydrochloride, ethanamine, 2,2'-dithiobis-, dihydrochloride, usaf cb-34, 2,2'-disulfanediyldiethanamine dihydrochloride, 2-aminoethyl disulfide dihydrochloride, bis 2-aminoethyl disulfide dihydrochloride, cystamine 2hcl, 2,2'-dithiodi ethylammonium dichloride, decarboxycystine dihydrochloride
PubChem CID 5941
IUPAC Name 2-(2-aminoethyldisulfanyl)ethanamine;dihydrochloride
SMILES C(CSSCCN)N.Cl.Cl

Specifications

Color White
Assay Percent Range min. 97.0%
Quantity 1 g
Formula Weight 225.2
Physical Form Powder
Chemical Name or Material Cystamine dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.