Learn More
Cystatin E/M Mouse anti-Human, Clone: 211515, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody has been used in 1 publication
Brand:  R&D Systems MAB1286-SP
Encompass
Product Code. 15753994
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Cystatin E/M/CST6 Monoclonal specifically detects Cystatin E/M/CST6 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Neutralization.