missing translation for 'onlineSavingsMsg'
Learn More

Cysteamine Hydrochloride, TRC

Product Code. 17931315
Change view
Click to view available options
Quantity:
50 mg
10 g
25 g
Unit Size:
10g
25g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17931315 50 mg 50mg
17964565 25 g 25g
17950815 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17931315 Supplier Toronto Research Chemicals Supplier No. TRCC99200050MG
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 156-57-0
Molecular Formula C2 H7 N S . Cl H
Molecular Weight (g/mol) 113.61
Synonym Ethanethiol, 2-amino-, hydrochloride (1:1), Ethanethiol, 2-amino-, hydrochloride (8CI,9CI), 1-Amino-2-mercaptoethane hydrochloride, 2-Aminoethanethiol hydrochloride, 2-Aminoethanthiol hydrochloride, 2-Mercaptoethanamine hydrochloride, 2-Mercaptoethanaminium chloride, 2-Mercaptoethylamine hydrochloride, 2-Mercaptoethylammonium chloride, 2-Thioethylamine hydrochloride, CI 9148, CT 2000, Cysteamine chlorohydrate, Cysteamine hydrochloride, Cysteaminium chloride, Cysteinamine hydrochloride, Lynovex, Mercamine hydrochloride, Merkamin hydrochloride, β-Mercaptoethylamine hydrochloride
IUPAC Name 2-aminoethanethiol;hydrochloride
SMILES Cl.NCCS

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C2H7NS.ClH/c3-1-2-4;/h4H,1-3H2;1H
Formula Weight 113.0066
Chemical Name or Material Cysteamine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.