missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Cytidine

Product Code. 16442069
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16442069 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16442069 Supplier MedChemExpress Supplier No. HYB0158
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Cytidine is a pyrimidine nucleoside and acts as a component of RNA. Cytidine is a precursor of uridine. Cytidine controls neuronal-glial glutamate cycling, affecting cerebral phospholipid metabolism, catecholamine synthesis, and mitochondrial function.

Specifications

Molecular Weight (g/mol) 243.22 g/mol
Product Type Natural Products
Content And Storage Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Shipping Condition Room temperature
Purity 98.78%
Quantity 1 g
Solubility Information H2O : ≥ 50 mg/mL (205.58 mM) ∣DMSO : 50 mg/mL (205.58 mM∣Need ultrasonic)
For Use With (Application) Cancer-programmed cell death
CAS 65-46-3
Synonym Cytosine β-D-riboside∣ Cytosine-1-β-D-ribofuranoside
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.