missing translation for 'onlineSavingsMsg'
Learn More

Cytidine-5-diphosphate Sodium Salt, TRC

Product Code. 30137569
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137569 5 g 5g
30006370 1 g 1g
30004911 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137569 Supplier Toronto Research Chemicals Supplier No. TRCC9983235G
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51855-98-2
Molecular Formula C9H15N3O11P2 . xNa
Molecular Weight (g/mol) 447.14
Synonym CDP Sodium Salt
IUPAC Name disodium;[[(2R,3S,4R,5R)-5-(4-amino-2-oxopyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-oxidophosphoryl] hydrogen phosphate
SMILES [Na+].[Na+].NC1=NC(=O)N(C=C1)[C@@H]2O[C@H](COP(=O)([O-])OP(=O)(O)[O-])[C@@H](O)[C@H]2O

Specifications

Quantity 5 g
InChI Formula InChI=1S/C9H15N3O11P2.2Na/c10-5-1-2-12(9(15)11-5)8-7(14)6(13)4(22-8)3-21-25(19,20)23-24(16,17)18;;/h1-2,4,6-8,13-14H,3H2,(H,19,20)(H2,10,11,15)(H2,16,17,18);;/q;2*+1/p-2/t4-,6-,7-,8-;;/m1../s1
Formula Weight 425.0001
Chemical Name or Material Cytidine-5-diphosphate Sodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.