missing translation for 'onlineSavingsMsg'
Learn More

Cytidine 5'-Monophosphate, TRC

Product Code. 30006165
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30006165 1 g 1g
30005129 5 g 5g
17988584 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30006165 Supplier Toronto Research Chemicals Supplier No. TRCC9983051G
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63-37-6
Molecular Formula C9 H14 N3 O8 P
Molecular Weight (g/mol) 323.2
Synonym 5'-Cytidylic acid, 5'-CMP, CMP, CMP (nucleotide), Cytidine 5'-(dihydrogen phosphate), Cytidine 5'-monophosphate, Cytidine 5'-monophosphoric acid, Cytidine 5'-phosphate, Cytidine 5'-phosphoric acid, Cytidine monophosphate, Cytidine, mono(dihydrogen phosphate) (ester), Cytidylic acid, Cytosine 5'-monophosphate
IUPAC Name [(2R,3S,4R,5R)-5-(4-amino-2-oxopyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl dihydrogen phosphate
SMILES NC1=NC(=O)N(C=C1)[C@@H]2O[C@H](COP(=O)(O)O)[C@@H](O)[C@H]2O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C9H14N3O8P/c10-5-1-2-12(9(15)11-5)8-7(14)6(13)4(20-8)3-19-21(16,17)18/h1-2,4,6-8,13-14H,3H2,(H2,10,11,15)(H2,16,17,18)/t4-,6-,7-,8-/m1/s1
Formula Weight 323.0519
Percent Purity >95%
Chemical Name or Material Cytidine-5'-monophosphate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.