missing translation for 'onlineSavingsMsg'
Learn More

Cytidine, free base, 99.8%, MP Biomedicals™

Product Code. 11473420 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
10 g
100 g
Unit Size:
100g
10g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11473420 5 g 5g
11483420 10 g 10g
11493420 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11473420 Supplier MP Biomedicals Supplier No. 0210145905
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 65-46-3
Molecular Formula C9H13N3O5
Molecular Weight (g/mol) 243.22
InChI Key UHDGCWIWMRVCDJ-UHFFFAOYNA-N
Synonym cytidine, cytosine riboside, 1-beta-d-ribofuranosylcytosine, 1beta-ribofuranosylcytosine, 4-amino-1-beta-d-ribofuranosyl-2 1h-pyrimidinone, beta-d-ribofuranoside, cytosine-1, 1-beta-ribofuranosylcytosine, cytidin, zytidin, 4-amino-1beta-d-ribofuranosyl-2 1h-pyrimidinone
PubChem CID 6175
ChEBI CHEBI:17562
IUPAC Name 4-amino-1-[3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2-dihydropyrimidin-2-one
SMILES NC1=NC(=O)N(C=C1)C1OC(CO)C(O)C1O

Specifications

Color White
Assay Percent Range 99.8%
Quantity 5 g
Formula Weight 243.22
Physical Form Crystalline Powder
Chemical Name or Material Cytidine, free base
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.