missing translation for 'onlineSavingsMsg'
Learn More

Cytidine, MP Biomedicals™

Product Code. 11435132 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11435132 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11435132 Supplier MP Biomedicals Supplier No. 0219465101
 Restricted Item
Estimated Shipment: 18-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Composed of the base cytosine linked to the five-carbon sugar d-ribose. Cytidine, MP Biomedicals is a pyrimidine nucleoside with a variety of analogs that display potentially useful pharmacology.

Cytidine, when intraperitoneal administered in combination with choline, improves nerve regeneration and functional recovery in a rat model of sciatic nerve injury. It is also used as an ex vivo drug selectable marker in gene-modified primary bone marrow stromal cells for cytidine deaminase assay.

Specifications

CAS 65-46-3
Absorbance λ max (water): 270-272nm
Melting Point 208°C to 218°C
Molecular Formula C9H13N3O5
Quantity 1 g
Synonym cytidine, cytosine riboside, 1-beta-d-ribofuranosylcytosine, 1beta-ribofuranosylcytosine, 4-amino-1-beta-d-ribofuranosyl-2 1h-pyrimidinone, beta-d-ribofuranoside, cytosine-1, 1-beta-ribofuranosylcytosine, cytidin, zytidin, 4-amino-1beta-d-ribofuranosyl-2 1h-pyrimidinone
Solubility Information Soluble in water (50mg/mL, clear colorless to faint yellow solution).
InChI Key UHDGCWIWMRVCDJ-UHFFFAOYNA-N
SMILES NC1=NC(=O)N(C=C1)C1OC(CO)C(O)C1O
IUPAC Name 4-amino-1-[3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2-dihydropyrimidin-2-one
Molecular Weight (g/mol) 243.22
PubChem CID 6175
ChEBI CHEBI:17562
Formula Weight 243.2
Grade Cell Culture Reagent
Physical Form Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.