missing translation for 'onlineSavingsMsg'
Learn More

Cytidine, TRC

Product Code. 30132809
Change view
Click to view available options
Quantity:
50 mg
1 g
5 g
Unit Size:
1g
50mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30132809 5 g 5g
30004569 50 mg 50mg
30003818 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132809 Supplier Toronto Research Chemicals Supplier No. TRCC9983005G
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65-46-3
Molecular Formula C9 H13 N3 O5
Molecular Weight (g/mol) 243.22
Synonym Cytidine, Cytosine, 1-β-D-ribosyl- (6CI), 1-(β-D-Ribofuranosyl)-2-oxo-4-amino-1,2-dihydro-1,3-diazine, 1-β-D-Ribofuranosylcytosine, 2(1H)-Pyrimidinone, 4-amino-1-β-D-ribofuranosyl-, 4-Amino-1-β-D-ribofuranosyl-2(1H)-pyrimidinone, Cytosine riboside, NSC 20258, β-D-Ribofuranoside, cytosine-1
IUPAC Name 4-amino-1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one
SMILES NC1=NC(=O)N(C=C1)[C@@H]2O[C@H](CO)[C@@H](O)[C@H]2O

Specifications

Quantity 5 g
InChI Formula InChI=1S/C9H13N3O5/c10-5-1-2-12(9(16)11-5)8-7(15)6(14)4(3-13)17-8/h1-2,4,6-8,13-15H,3H2,(H2,10,11,16)/t4-,6-,7-,8-/m1/s1
Formula Weight 243.0855
Chemical Name or Material Cytidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.